AMBH324A57B0 Ambeed, Inc.

Boc-Ala-D-Glu(OBzl)-OH

  • MDL number MFCD28978616
  • Cas Number : 53759-36-7
  • Molecular Weight : 408.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/53759-36-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/53759-36-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart