AMBH324A57C5 Ambeed, Inc.

(S)-Methyl 3-hydroxy-2-(tritylamino)propanoate

  • MDL number MFCD00192570
  • Cas Number : 4465-44-5
  • Molecular Weight : 361.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/4465-44-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/4465-44-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart