AMBH324A57C6 Ambeed, Inc.

2-(Piperazin-1-yl)quinoline maleate

  • MDL number MFCD00133796
  • Cas Number : 5786-68-5
  • Molecular Weight : 329.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/5786-68-5.html

Safety Information

signalword Danger
pcodes P301+P310
MSDS https://www.ambeed.com/products/5786-68-5.html
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart