AMBH324A57D7 Ambeed, Inc.

Methyl-5-norbornene-2,3-dicarboxylic Anhydride (mixture of isomers)

  • MDL number MFCD00167036
  • Cas Number : 25134-21-8
  • Molecular Weight : 178.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/25134-21-8.html

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/25134-21-8.html
pictograms GHS05,GHS07,GHS08
hstatements H302-H314-H317-H334
Availability details for
Quantity
  • Availability
  • Add to Cart