AMBH324A57DB Ambeed, Inc.

Resveratrol

  • MDL number MFCD00133799
  • Cas Number : 501-36-0
  • Molecular Weight : 228.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/501-36-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/501-36-0.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart