AMBH324A57DD Ambeed, Inc.

(S)-Methyl 4-(piperidin-2-yl)benzoate hydrochloride

  • MDL number MFCD12910878
  • Cas Number : 1391547-09-3
  • Molecular Weight : 255.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1391547-09-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1391547-09-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart