AMBH324A57DE Ambeed, Inc.

Tylosin tartrate

  • MDL number MFCD07783824
  • Cas Number : 74610-55-2
  • Molecular Weight : 1052.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/74610-55-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/74610-55-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart