AMBH324A57E5 Ambeed, Inc.

Sodium (3S,4R,5R)-6-((hydrogenphosphonato)oxy)-3,4,5-trihydroxy-2-oxohexyl phosphate octahydrate

  • MDL number MFCD00149340
  • Cas Number : 81028-91-3
  • Molecular Weight : 550.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/81028-91-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/81028-91-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart