AMBH324A57F0 Ambeed, Inc.

(R)-N-Methyl-1-phenylethanamine hydrochloride

  • MDL number N/A
  • Cas Number : 1096105-18-8
  • Molecular Weight : 171.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1096105-18-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1096105-18-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart