AMBH324A57F5 Ambeed, Inc.

(3R,4S,5S)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxydihydrofuran-2(3H)-one

  • MDL number MFCD00005391
  • Cas Number : 6322-07-2
  • Molecular Weight : 178.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/6322-07-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6322-07-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart