AMBH324A57F7 Ambeed, Inc.

(S)-Methyl 3-amino-2-((tert-butoxycarbonyl)amino)propanoate hydrochloride

  • MDL number MFCD04973085
  • Cas Number : 181228-33-1
  • Molecular Weight : 254.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/181228-33-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/181228-33-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart