AMBH324A57F8 Ambeed, Inc.

Gemcitabine

  • MDL number MFCD00869720
  • Cas Number : 95058-81-4
  • Molecular Weight : 263.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/95058-81-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/95058-81-4.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart