AMBH324A57FC Ambeed, Inc.

Fmoc-Lys(ivDde)-OH

  • MDL number MFCD01631658
  • Cas Number : 204777-78-6
  • Molecular Weight : 574.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/204777-78-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/204777-78-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart