AMBH324A580D Ambeed, Inc.

H-Lys(Z)-NH2.HCl

  • MDL number MFCD00035520
  • Cas Number : 58117-53-6
  • Molecular Weight : 315.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/58117-53-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/58117-53-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart