AMBH324A581B Ambeed, Inc.

(2R,3R)-2,3-Bis(pivaloyloxy)succinic acid

  • MDL number MFCD00066535
  • Cas Number : 65259-81-6
  • Molecular Weight : 318.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/65259-81-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/65259-81-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart