AMBH324A5832 Ambeed, Inc.

NSC 632839 Hydrochloride

  • MDL number MFCD09753281
  • Cas Number : 157654-67-6
  • Molecular Weight : 339.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/157654-67-6.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/157654-67-6.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart