AMBH324A5835 Ambeed, Inc.

4-Ethoxy-1,1,1-trifluorobut-3-en-2-one

  • MDL number MFCD00192131
  • Cas Number : 17129-06-5
  • Molecular Weight : 168.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/17129-06-5.html

Safety Information

signalword Danger
pcodes P201-P308+P313-P403+P235
MSDS https://www.ambeed.com/products/17129-06-5.html
pictograms GHS02,GHS08
hstatements H226-H340-H351
Availability details for
Quantity
  • Availability
  • Add to Cart