AMBH324A5837 Ambeed, Inc.

Schizandrin

  • MDL number MFCD00905761
  • Cas Number : 7432-28-2
  • Molecular Weight : 432.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/7432-28-2.html

Safety Information

signalword Danger
pcodes P301+P312+P330
MSDS https://www.ambeed.com/products/7432-28-2.html
pictograms GHS07,GHS09
hstatements H302-H400
Availability details for
Quantity
  • Availability
  • Add to Cart