AMBH324A5844 Ambeed, Inc.

NPS-2143 hydrochloride

  • MDL number N/A
  • Cas Number : 324523-20-8
  • Molecular Weight : 445.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/324523-20-8.html

Safety Information

signalword Danger
pcodes P273-P280-P501
MSDS https://www.ambeed.com/products/324523-20-8.html
pictograms GHS07,GHS09
hstatements H302+H312+H332-H410
Availability details for
Quantity
  • Availability
  • Add to Cart