AMBH324A5856 Ambeed, Inc.

(S)-Methyl 2-amino-3-(3,4-dimethoxyphenyl)propanoate hydrochloride

  • MDL number MFCD07778593
  • Cas Number : 70494-48-3
  • Molecular Weight : 275.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/70494-48-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/70494-48-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart