AMBH324A5857 Ambeed, Inc.

Boc-N-Me-Tyr-OH.DCHA

  • MDL number MFCD00237569
  • Cas Number : 95105-25-2
  • Molecular Weight : 476.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/95105-25-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/95105-25-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart