AMBH324A585B Ambeed, Inc.

FX1

  • MDL number N/A
  • Cas Number : 1426138-42-2
  • Molecular Weight : 368.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1426138-42-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1426138-42-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart