AMBH324A5871 Ambeed, Inc.

(E)-2-Phenylethene-1-sulfonyl chloride

  • MDL number MFCD00007456
  • Cas Number : 52147-97-4
  • Molecular Weight : 202.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/52147-97-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/52147-97-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart