AMBH324A587D Ambeed, Inc.

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,4,5-trifluorophenyl)propanoic acid

  • MDL number MFCD00797583
  • Cas Number : 205526-30-3
  • Molecular Weight : 441.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/205526-30-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/205526-30-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart