AMBH324A5888 Ambeed, Inc.

((1S,4R)-4-(2-Amino-6-chloro-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hydrochloride

  • MDL number MFCD12546177
  • Cas Number : 172015-79-1
  • Molecular Weight : 302.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/172015-79-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/172015-79-1.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart