AMBH324A588B Ambeed, Inc.

Penicillin G sodium salt

  • MDL number MFCD00069666
  • Cas Number : 69-57-8
  • Molecular Weight : 356.3700

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 1500
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/69-57-8.html

Safety Information

signalword Warning
pcodes P261-P280
MSDS https://www.ambeed.com/products/69-57-8.html
pictograms GHS07
hstatements H317
Availability details for
Quantity
  • Availability
  • Add to Cart