AMBH324A5892 Ambeed, Inc.

Furazolidone

  • MDL number MFCD00010550
  • Cas Number : 67-45-8
  • Molecular Weight : 225.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/67-45-8.html

Safety Information

signalword Danger
pcodes P281
MSDS https://www.ambeed.com/products/67-45-8.html
pictograms GHS08
hstatements H361
Availability details for
Quantity
  • Availability
  • Add to Cart