AMBH324A589A Ambeed, Inc.

(S)-Ethyl 2-(3-((2-isopropylthiazol-4-yl)methyl)-3-methylureido)-4-morpholinobutanoate oxalate

  • MDL number MFCD28977898
  • Cas Number : 1247119-36-3
  • Molecular Weight : 502.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1247119-36-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1247119-36-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart