AMBH324A58B1 Ambeed, Inc.

Boc-Asp(OtBu)-ONp

  • MDL number MFCD00076913
  • Cas Number : 29365-05-7
  • Molecular Weight : 410.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/29365-05-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/29365-05-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart