AMBH324A58B7 Ambeed, Inc.

(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate

  • MDL number MFCD00149364
  • Cas Number : 16373-93-6
  • Molecular Weight : 269.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/16373-93-6.html

Safety Information

signalword Warning
pcodes P301+P312
MSDS https://www.ambeed.com/products/16373-93-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart