AMBH324A58BF Ambeed, Inc.

Penicillin V Potassium

  • MDL number MFCD00051771
  • Cas Number : 132-98-9
  • Molecular Weight : 388.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/132-98-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/132-98-9.html
pictograms GHS07
hstatements H302-H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart