AMBH324A58CE Ambeed, Inc.

Salermide

  • MDL number MFCD12912446
  • Cas Number : 1105698-15-4
  • Molecular Weight : 394.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1105698-15-4.html

Safety Information

signalword Warning
pcodes P273-P280
MSDS https://www.ambeed.com/products/1105698-15-4.html
pictograms GHS07
hstatements H317-H412
Availability details for
Quantity
  • Availability
  • Add to Cart