AMBH324A58F0 Ambeed, Inc.

N-Methyl-L-proline Monohydrate

  • MDL number MFCD00149962
  • Cas Number : 199917-42-5
  • Molecular Weight : 147.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/199917-42-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/199917-42-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart