AMBH324A58F8 Ambeed, Inc.

(R)-2-Hydroxy-1,2,2-triphenylethyl acetate

  • MDL number MFCD00066242
  • Cas Number : 95061-47-5
  • Molecular Weight : 332.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/95061-47-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/95061-47-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart