AMBH324A58FA Ambeed, Inc.

DL-2,3-Dihydroxy-1,4-butanedithiol

  • MDL number MFCD00004877
  • Cas Number : 3483-12-3
  • Molecular Weight : 154.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/3483-12-3.html

Safety Information

signalword Warning
pcodes P273
MSDS https://www.ambeed.com/products/3483-12-3.html
pictograms GHS07
hstatements H302-H412
Availability details for
Quantity
  • Availability
  • Add to Cart