AMBH324A58FE Ambeed, Inc.

(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol

  • MDL number MFCD01318247
  • Cas Number : 86695-06-9
  • Molecular Weight : 255.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/86695-06-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/86695-06-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart