AMBH324A591C Ambeed, Inc.

(R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid hydrate

  • MDL number N/A
  • Cas Number : 350230-37-4
  • Molecular Weight : 223.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/350230-37-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/350230-37-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart