AMBH324A5923 Ambeed, Inc.

Ethyl cinnamate

  • MDL number MFCD00009189
  • Cas Number : 103-36-6
  • Molecular Weight : 176.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/103-36-6.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/103-36-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart