AMBH324A592E Ambeed, Inc.

(2E,4E)-Hepta-2,4-dienal

  • MDL number MFCD00007005
  • Cas Number : 4313-03-5
  • Molecular Weight : 110.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/4313-03-5.html

Safety Information

signalword Danger
pcodes P280-P312
MSDS https://www.ambeed.com/products/4313-03-5.html
pictograms GHS06
hstatements H302-H311-H315
Availability details for
Quantity
  • Availability
  • Add to Cart