AMBH324A593A Ambeed, Inc.

Tirofiban hydrochloride monohydrate

  • MDL number MFCD07368623
  • Cas Number : 150915-40-5
  • Molecular Weight : 495.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/150915-40-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/150915-40-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart