AMBH324A593B Ambeed, Inc.

(1S,2R)-2-(Benzyloxymethyl)-1-hydroxy-3-cyclopentene

  • MDL number MFCD09263486
  • Cas Number : 110567-21-0
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/110567-21-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/110567-21-0.html
pictograms GHS07
hstatements H227-H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart