AMBH324A5948 Ambeed, Inc.

(S)-1-(Cyclopropylmethyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD18070744
  • Cas Number : 342793-01-5
  • Molecular Weight : 169.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/342793-01-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/342793-01-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart