AMBH324A594B Ambeed, Inc.

Sal003

  • MDL number N/A
  • Cas Number : 1164470-53-4
  • Molecular Weight : 463.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1164470-53-4.html

Safety Information

signalword Danger
pcodes P273
MSDS https://www.ambeed.com/products/1164470-53-4.html
pictograms GHS09
hstatements H410
Availability details for
Quantity
  • Availability
  • Add to Cart