AMBH324A5950 Ambeed, Inc.

Biapenem

  • MDL number MFCD00864863
  • Cas Number : 120410-24-4
  • Molecular Weight : 350.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/120410-24-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/120410-24-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart