AMBH324A5958 Ambeed, Inc.

Sodium (2R,3R,4S,5R,6S)-2,3,4,5,6,7-hexahydroxyheptanoate dihydrate

  • MDL number N/A
  • Cas Number : 10094-62-9
  • Molecular Weight : 284.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/10094-62-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/10094-62-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart