AMBH324A5969 Ambeed, Inc.

(E)-2,2'-(Fumaroylbis(oxy))bis(3,5-dibromobenzoic acid)

  • MDL number MFCD03547939
  • Cas Number : 71337-53-6
  • Molecular Weight : 671.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/71337-53-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/71337-53-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart