AMBH324A596D Ambeed, Inc.

lumateperone

  • MDL number MFCD28502161
  • Cas Number : 313368-91-1
  • Molecular Weight : 393.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/313368-91-1.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/313368-91-1.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart