AMBH324A5975 Ambeed, Inc.

Colimycin sulfate

  • MDL number MFCD00146495
  • Cas Number : 1264-72-8
  • Molecular Weight : 1267.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 19000
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1264-72-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1264-72-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart