AMBH324A597A Ambeed, Inc.

trans-4-Aminocyclohexanol

  • MDL number MFCD00067698
  • Cas Number : 27489-62-9
  • Molecular Weight : 115.1735

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/27489-62-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/27489-62-9.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart