AMBH324A598C Ambeed, Inc.

Sodium Diformylamide

  • MDL number MFCD00145302
  • Cas Number : 18197-26-7
  • Molecular Weight : 96.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/18197-26-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/18197-26-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart